missing translation for 'onlineSavingsMsg'
Learn More

Coomassie Brilliant Blue R-250, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0219068280

 View more versions of this product

Catalog No. ICN19068280


Only null left

Description

Description

Specifications

Specifications

Coomassie Brilliant Blue R-250
6104-59-2
C45H44N3NaO7S2
Brilliant Blue R, Acid Blue 83
[Na+].CCOC1=CC=C(NC2=CC=C(C=C2)[C+](C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C=C1
825.97
826
100 g
Protein stain in electrophoresis.
∽ 50%
MFCD00041762
HAEGGLCXLFWTBE-UHFFFAOYSA-M
sodium {4-[(4-ethoxyphenyl)amino]phenyl}bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)methylium
61365
Brown to Purple
Powder

Chemical Identifiers

6104-59-2
825.97
HAEGGLCXLFWTBE-UHFFFAOYSA-M
61365
[Na+].CCOC1=CC=C(NC2=CC=C(C=C2)[C+](C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C=C1
C45H44N3NaO7S2
MFCD00041762
Brilliant Blue R, Acid Blue 83
sodium {4-[(4-ethoxyphenyl)amino]phenyl}bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)methylium
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions