missing translation for 'onlineSavingsMsg'
Learn More

Coomassie Brilliant Blue R-250, MP Biomedicals™

Catalog No. p-4405561
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19068205 5 g
ICN19068225 25 g
ICN19068280 100 g
ICN19068290 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19068205 Supplier MP Biomedicals Inc Supplier No. 0219068205
Only null left

Chemical Identifiers

CAS 6104-59-2
Molecular Formula C45H44N3NaO7S2
Molecular Weight (g/mol) 825.97
MDL Number MFCD00041762
InChI Key HAEGGLCXLFWTBE-UHFFFAOYSA-M
Synonym Brilliant Blue R, Acid Blue 83
PubChem CID 61365
IUPAC Name sodium {4-[(4-ethoxyphenyl)amino]phenyl}bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)methylium
SMILES [Na+].CCOC1=CC=C(NC2=CC=C(C=C2)[C+](C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C=C1

Specifications

Assay Percent Range ∽ 50%
Formula Weight 826
Color Brown to Purple
Quantity 5 g
Physical Form Powder
Chemical Name or Material Coomassie Brilliant Blue R-250, Protein stain in electrophoresis.