Learn More
Corning™ CoolRack™ M-PF Conical Tube Modules
Ensures ±0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
$816.99 - $1,235.51
Specifications
Type Conical Tube Thermoconductive Module
View More Specs
Products 3
Catalog Number Mfr. No. Holds Design Price Quantity & Availability  
Catalog Number Mfr. No. Holds Design Price Quantity & Availability  
07210048
View Documents Promotion Details
Corning™
432048
30 x 1.5 mL Conical Tubes M30-PF
Case for $1,235.51
Only null left
07210046
View Documents Promotion Details
Corning™
432046
30 x 500μL Conical Tubes M30-PF
Case for $1,235.51
Only null left
07210047
View Documents Promotion Details
Corning™
432047
15 x 1.5 mL conical tubes M15-PF
Case for $816.99
Only null left
Description
Description
Corning CoolRack thermo-conductive microfuge tube modules eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources. Place the CoolRack module directly onto a temperature source between -196°C to >100°C and it will rapidly adapt to that temperature.
  • Ensures ± 0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
  • All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
  • Suggested applications include cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen.
Specifications
Specifications
Conical Tube Thermoconductive Module