missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ CoolRack™ M-PF Conical Tube Modules
Ensures ±0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
Supplier: Corning™ 432047
Description
Corning CoolRack thermo-conductive microfuge tube modules eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources. Place the CoolRack module directly onto a temperature source between -196°C to >100°C and it will rapidly adapt to that temperature.
- Ensures ± 0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
- All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
- Suggested applications include cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen.
Chemical Identifiers
| Conical Tube Thermoconductive Module |
Specifications
| 15 x 1.5 mL conical tubes | |
| M15-PF | |
| Conical Tube Thermoconductive Module |