Learn More
Corning™ CoolRack™ CF Cryogenic Vial and FACS Tube Modules
Ensures ±0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
$573.59 - $1,189.84
Specifications
Type Cryogenic Vial, FACS Tube Module
View More Specs
Products 4
Catalog Number Mfr. No. Holds Price Quantity & Availability  
Catalog Number Mfr. No. Holds Price Quantity & Availability  
07210049
View Documents Promotion Details
Corning™
432049
15 Cryogenic Vials or FACS Tubes
Case for $573.59
Only null left
07210050
View Documents Promotion Details
Corning™
432050
24 Cryogenic Vials or FACS Tubes
Case for $1,029.11
Only null left
07210051
View Documents Promotion Details
Corning™
432051
45 Cryogenic Vials or FACS Tubes
Case for $1,114.80
Only null left
07210052
View Documents Promotion Details
Corning™
432052
30 Cryogenic Vials or FACS Tubes
Case for $1,189.84
Only null left
Description
Description
Corning CoolRack thermo-conductive tube modules eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources. Place the CoolRack module directly onto a temperature source between -196°C to > 100°C and it will rapidly adapt to that temperature.
  • Ensure ± 0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
  • All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
  • Suggested applications include cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen.
Specifications
Specifications
Cryogenic Vial, FACS Tube Module