Learn More
Corning™ CoolRack™ CF Cryogenic Vial and FACS Tube Modules
Ensures ±0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
Supplier: Corning™ 432050
Description
Corning CoolRack thermo-conductive tube modules eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources. Place the CoolRack module directly onto a temperature source between -196°C to > 100°C and it will rapidly adapt to that temperature.
- Ensure ± 0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
- All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
- Suggested applications include cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen.
Chemical Identifiers
| Cryogenic Vial, FACS Tube Module |
Specifications
| 24 Cryogenic Vials or FACS Tubes | |
| XT CFT24, w/gripping wells for one-hand 1 vial opening/closing, SBS-compatible | |
| Cryogenic Vial, FACS Tube Module |