Learn More
Corning™ Extension Collar for CoolBox™ 2XT System
For use with CoolBox™ 2XT System. Corning™ Extension Collar is required when using taller CoolRack™ tube modules and tall tubes in CoolBox system.
$422.93
Specifications
No. per Pack 1/Pk.
No. per Case 1/Cs.
For Use With (Equipment) CoolBox 2XT System
View More Specs
Alert:
Lid closure not possible with addition of extension collar
Products 4
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
07210087
View Documents Promotion Details
Corning™
432087
Purple
Case for $422.93
Only null left
07210088
View Documents Promotion Details
Corning™
432088
Green
Case for $422.93
Only null left
07210089
View Documents Promotion Details
Corning™
432089
Orange
Case for $422.93
Only null left
07210090
View Documents Promotion Details
Corning™
432090
Pink
Case for $422.93
Only null left
Description
Description
For use with CoolBox XT and CoolBox 2XT to extend the height when using tall tube modules. The collar is magnetized and easily adheres to the unit base.
Specifications
Specifications
1/Pk.
1/Cs.
CoolBox 2XT System