Learn More
Corning™ CoolCell™ FTS30 Cell Freezing Vial Containers
Vials achieve a uniform and reproducible -1°C/minute freezing profile and thermal profiles are highly reproducible.
Supplier:  Corning™ 432007
 View more versions of this product
Catalog No. 07210007
Only null left
Description
Description
Corning CoolCell, in combination with a -80°C freezer, will provide alcohol-free freezing at the rate of -1°C/minute that is ideal for cryopreservation of most cells and cell lines. Using a combination of uniform-density cross-linked polyethylene foam, a solid state core, and radial vial symmetry, freezing profiles are consistent and reproducible. The foam is non-absorbent and will impose negligible change in the freezer environment; thereby protecting nearby frozen samples. The removal of vials is accomplished by tipping the device upside down so the vials slide out.
Features: Unique features of controlled-rate freezing with Corning™ CoolCell™ include:
  • Ease of use
  • Alcohol and fluid-free freezing
  • Lower cost of use than alcohol-based devices
  • High cell recovery and cell viability
  • Reproducibility
  • Simple, consistent way to standardize controlled-rate freezing
Specifications
Chemical Identifiers
Polyethylene Foam
Freezer Box
1/Pk., 1/Cs.
16.5 cm
12.7 mm
Round
30 (Standard 1 to 2 mL Cryogenic Vials)
16.5 x 11.5 cm
11.5 cm
1 kg
Specifications
Orange
Round
30 (Standard 1 to 2 mL Cryogenic Vials)
16.5 x 11.5 cm
11.5 cm
1 kg
Polyethylene Foam
Freezer Box
1/Pk., 1/Cs.
16.5 cm
12.7 mm
Product Suggestions
Product Suggestions