missing translation for 'onlineSavingsMsg'
Learn More

Corning™ CoolCell™ FTS30 Cell Freezing Vial Containers

Catalog No. p-6936054
Change view
Click to view available options
Color:
Green
Orange
Pink
Purple
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07210006 Purple
07210007 Orange
07210008 Green
07210009 Pink
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 07210006 Supplier Corning™ Supplier No. 432006

Vials achieve a uniform and reproducible -1°C/minute freezing profile and thermal profiles are highly reproducible.

Corning CoolCell, in combination with a -80°C freezer, will provide alcohol-free freezing at the rate of -1°C/minute that is ideal for cryopreservation of most cells and cell lines. Using a combination of uniform-density cross-linked polyethylene foam, a solid state core, and radial vial symmetry, freezing profiles are consistent and reproducible. The foam is non-absorbent and will impose negligible change in the freezer environment; thereby protecting nearby frozen samples. The removal of vials is accomplished by tipping the device upside down so the vials slide out.

Features: Unique features of controlled-rate freezing with Corning™ CoolCell™ include:

  • Ease of use
  • Alcohol and fluid-free freezing
  • Lower cost of use than alcohol-based devices
  • High cell recovery and cell viability
  • Reproducibility
  • Simple, consistent way to standardize controlled-rate freezing

Specifications

Color Purple
Material Polyethylene Foam
Shape Round
Type Freezer Box
Holds 30 (Standard 1 to 2 mL Cryogenic Vials)
Quantity 1/Pk., 1/Cs.
Dimensions (L x W) 16.5 x 11.5 cm
Length (Metric) 16.5 cm
Width (Metric) 11.5 cm
Diameter (Metric) Well 12.7 mm
Weight (Metric) 1 kg
Show More Show Less