Learn More
Corning™ Extension Collar for CoolBox™ XT System
For use with CoolBox™ XT System. Corning™ Extension Collar is required when using taller CoolRack™ tube modules and tall tubes in CoolBox system.
Supplier:  Corning™ 432084
 View more versions of this product
Catalog No. 07210084
Only null left
Alert:
Lid closure not possible with addition of extension collar
Description
Description
For use with CoolBox XT and CoolBox 2XT to extend the height when using tall tube modules. The collar is magnetized and easily adheres to the unit base.
Specifications
Chemical Identifiers
Extension Collar
1/Pk.
20.0 x 16.0 x 7.0 cm (7.8 x 6.2 x 2.7 in.) Interior: 14.0 x 9.4 x 7.0 cm (5.5 x 3.7 x 2.7 in.)
Specifications
Green
Extension Collar
CoolBox XT and CoolBox 2XT
20.0 x 16.0 x 7.0 cm (7.8 x 6.2 x 2.7 in.) Interior: 14.0 x 9.4 x 7.0 cm (5.5 x 3.7 x 2.7 in.)
20 cm
16 cm
7 cm
Square
Extend the Height When Using Tall Tube Modules
1/Pk.
7.8 in.
6.2 in.
2.75 in.
Product Suggestions
Product Suggestions