Learn More
Fisherbrand™ Controlled Environment Boot Cover
Fisherbrand high top anti-skid boot covers fit over the calf with an elastic top to provide a comfortable fit and to prevent the boot cover from sliding down. Sole material is clean and durable. Upper material is soft and strong for all day comfort.
Supplier:  Fisherbrand™ BTA1613BFS
 View more versions of this product
Catalog No. 19-165-013XL
Only null left
Explore available promotions
Description
Description
  • Inherent non-skid physical properties
  • Strong enough to withstand even the most abrasive floor surfaces
  • Fabric provides barrier protection and exhibits low particle generation
  • High COF fit
Specifications
Specifications
X-Large
Yes
Single-use
Elastic Top and Ankles
Serged
200/Cs.
15 in.
Abrasion
Class 100 (ISO 5)
Polyethylene, Polypropylene
White
Non-sterile
Anti-skid
Controlled and Critical Environments
Double Bagged
18 in.
Controlled Environments Boot Covers
Product Suggestions
Product Suggestions
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'