missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Control DNA (from CEPH Individual 1347-02)

Catalog No. 403062
Encompass
Change view
Click to view available options
Quantity:
180 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
403062 180 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 403062 Supplier Applied Biosystems™ Supplier No. 403062

Includes

Contains control DNA in Tris buffer, pH 8.0, and EDTA.

The control DNA can be used with linkage mapping sets MD10, HD5, and LD20.

  • Functions as a control template to monitor PCR amplification
  • Serves as sizing reference for controlling gel-to-gel or capillary-to-capillary variation
  • Aids in the comparison of alleles sizes from different gels (allele binning)
  • Enables correlation with allele frequency data from external sources such as others databases and laboratories

Specifications

Content And Storage This product contains control DNA in Tris buffer, pH 8.0, and EDTA.
Format Tube
For Use With (Application) Sequencing
Includes Control Template(s)
No. of Reactions 150
Product Type Control DNA
Quantity 180 μL
Shipping Condition Dry Ice
Species Human

For Research Use Only. Not for use in diagnostic procedures.