missing translation for 'onlineSavingsMsg'
Learn More

Decon™ Contrex™ EZ Powdered Enzymatic Detergent

Formulated for removal of protein and fat contamination

$115.83 - $684.87

Specifications

Product Type Detergent
Color-coded No
Form Powder
pH 9.5 to 11
Scent Regular Scent
View More Specs

Products 2
Catalog Number Mfr. No. Volume (English) Price Quantity  
Catalog Number Mfr. No. Volume (English) Price Quantity  
04-358-23
View Documents
Decon™
5404
4 lb.
Each for $115.83
Case of 9 Each for $1,042.47
Only null left
 
04-358-24
View Documents
Decon™
5425
25 lb.
Each for $684.87
Only null left
 
Description

Description

Ideal for use in manual or soak cleaning of labware and equipment.

  • Powdered
  • Free-rinsing and disposable straight to drain
  • Enzymatic action
  • Environmentally Friendly: Item contains environmental attributes or features (e.g. water conserving, lead-free, etc.) as described in the item detail
  • Recycled: Item contains recycled content as described in the item detail
  • Non-toxic: Item is non-toxic
Specifications

Specifications

Detergent
Powder
Regular Scent
White
For use in manual or soak cleaning of labware and equipment contaminated with proteins and fats
No
9.5 to 11
Non-sterile
29 CFR1910.1200
Videos
SDS
Documents

Documents