Learn More
Contec™ Edgeless™ Mop System
GREENER_CHOICE
Clean in controlled environments and cleanrooms. The Edgeless™ Mop System is constructed from a continuous tube of 100% polyester knitted fabric. The mop heads help eliminate contamination from breakage and deterioration of traditional string mops.
Supplier:  Contec™ ELMOPTGY
 View more versions of this product
Catalog No. 19033851
Only null left
Description
Description
  • Designed for wet mopping in controlled environments and cleanrooms
  • Ideal for use in sterile environments
  • Mop heads are constructed from a continuous tube of 100% polyester knitted fabric, looped to form strands that encapsulate liquids for maximum absorption
  • Mop heads are free of any cellulosic contaminants
  • Textured mop heads feature ribbed, twisted strands for 60% more sorbent capacity than standard Edgeless mops
  • Textured knitted polyester tube mop heads help eliminate contamination from breakage and deterioration of traditional string mops
  • Mop heads are laundered and packaged in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom
Specifications
Chemical Identifiers
Non-sterile
Autoclavable
Limited-use
Polyester
Specifications
Cleanroom Mop
Gray, Black (Tail Band)
Autoclavable
20/Cs.
Contec Edgeless Mopping System
Designed for use with aqueous solutions containing cleaning agents and disinfectants; compatible with wide range of chemicals
Enhanced with Textured Headband
Non-sterile
Limited-use
Textured Headband
Cleanroom Laundered Enhanced Mop Head with Textured Headband
Polyester
Product Suggestions
Product Suggestions