missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Container Label: LACQUER THINNER

Catalog No. 19107295
Change view
Click to view available options
Adhesive Type:
Acrylic, Permanent, Cold-temperature
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Adhesive Type
19107295 Acrylic, Permanent, Cold-temperature
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19107295 Supplier Brady™ Supplier No. 60251

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Identify the contents of various containers with easy visibility. Brady™ Container Labels are made from laminated polyester and display text in red on yellow.

  • Pre-printed labels to identify container contents
  • Self-adhesive polyester labels made from laminated polyester
  • Chemically resistant for longer life

Specifications

Description Abrasion and chemical resistant material
Adhesive Type Acrylic, Permanent, Cold-temperature
Color Red, Yellow
Format Label
Material Laminated Polyester
Product Type Container Label
Display Text LACQUER THINNER
Quantity 25/Pk.
Shape Rectangle
Size (English) 7 x 3 in. (W x H)
Thickness (English) 0.01 in.
Width (English) 7 in.
Show More Show Less