missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Contactin 4 Polyclonal Antibody

Sheep Polyclonal Antibody
Supplier: Invitrogen™ PA5149014
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
The protein encoded by this gene is a member of the immunoglobulin superfamily. It is a glycosylphosphatidylinositol (GPI)-anchored neuronal membrane protein that functions as a cell adhesion molecule. It may play a role in the formation of axon connections in the developing nervous system. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.Chemical Identifiers
| Contactin 4 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Lyophilized | |
| IgG | |
| Mouse | |
| CNTN4 | |
| 9630050B05; Axcam; axonal-associated cell adhesion molecule; Big-2; brain-derived immunoglobulin superfamily molecule; brain-derived immunoglobulin superfamily protein 2; CNTN4; CNTN4A; contactin 4; contactin-4; Kiaa3024; MGC33615; neural cell adhesion protein BIG-2; SCA16 | |
| CNTN4 | |
| Primary | |
| Antigen affinity chromatography |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Sheep | |
| RUO | |
| PBS with 5% trehalose and no preservative | |
| Q69Z26 | |
| 269784 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse Contactin-4, Asp19-Gly999 (Ile64Leu). | |
| Antibody |
Specifications
| Contactin 4 | |
| Polyclonal | |
| Unconjugated | |
| CNTN4 | |
| 9630050B05; Axcam; axonal-associated cell adhesion molecule; Big-2; brain-derived immunoglobulin superfamily molecule; brain-derived immunoglobulin superfamily protein 2; CNTN4; CNTN4A; contactin 4; contactin-4; Kiaa3024; MGC33615; neural cell adhesion protein BIG-2; SCA16 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 269784 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and no preservative | |
| Q69Z26 | |
| CNTN4 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse Contactin-4, Asp19-Gly999 (Ile64Leu). | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |