missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Connexin 32 Recombinant Rabbit Monoclonal Antibody (23GB5895)
GREENER_CHOICE

Catalog No. MA552942
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552942 100 μL
MA552943 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552942 Supplier Invitrogen™ Supplier No. MA552942
Only null left

Rabbit Recombinant Monoclonal Antibody

Connexin 32 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

This gene encodes a member of the gap junction protein family. The gap junction proteins are membrane-spanning proteins that assemble to form gap junction channels that facilitate the transfer of ions and small molecules between cells. According to sequence similarities at the nucleotide and amino acid levels, the gap junction proteins are divided into two categories, alpha and beta. Mutations in this gene cause X-linked Charcot-Marie-Tooth disease, an inherited peripheral neuropathy. Alternatively spliced transcript variants encoding the same protein have been found for this gene.

Specifications

Antigen Connexin 32
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB5895
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GJB1
Gene Accession No. P08034
Gene Alias CMTX; CMTX1; Cnx32; connexin 32; connexin-32; CX32; Cxn-32; GAP junction 28 kDa liver protein; gap junction beta-1 protein; gap junction membrane channel protein beta 1; gap junction protein beta 1; gap junction protein, beta 1; gap junction protein, beta 1, 32kDa; Gjb1
Gene Symbols GJB1
Host Species Rabbit
Immunogen A synthesized peptide derived from human GJB1 (50-84AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2705
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less