missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Connector Cables for Automated Thermal Cycler (ATC)

Catalog No. A31483
Encompass
Change view
Click to view available options
Format:
1-m connector cable (flexible)
3-m connector cable (flexible)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
A31483 3-m connector cable (flexible)
A31482 1-m connector cable (flexible)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A31483 Supplier Applied Biosystems™ Supplier No. A31483
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used to connect the Automated Thermal Cycler (ATC) block to the power box, this connector is available in a rigid 10-cm, flexible 1-m, or flexible 3-m cable format.

For customers seeking to automate their PCR workflow, the Applied Biosystems Automated Thermal Cycler (ATC) provides the reliability and performance you've come to expect from Applied Biosystems instruments. Small, easy-to-integrate, with a compatible format, our Automated Thermal Cycler enables hands-free PCR results.

In order to better align with lab design and demands, a longer or shorter cable may be utilized to optimize workflow.

Specifications

Format 3-m connector cable (flexible)
Thermal Accuracy ±0.25°C (35°C to 99.9°C)
Temperature Range (Metric) 0.0 to 105.0°C
Content And Storage
  • ATC block module
  • ATC control module
  • Dell E6540 RC Laptop computer
  • 1 meter connection cable
  • CAT 5 ethernet cable – Store at room temp
For Use With (Equipment) Automated Thermal Cycler (ATC)
Thermal Uniformity <0.5 °C (20 s after reaching 95°C)
Dimensions 17.9 x 31.7 x 13.3 cm (Block Module)
Electrical Requirements Connectivity-LAN
Product Type Connector Cable
Warranty 27 Month Warranty
For Use With (Application) Applied Biosystems Automated Thermal Cyclers
Description Cabling Option - 3MPlate Format - 96-wellData Connectivity - LANBlock Module Dimensions - 17.9 x 31.7 x 13.3 cmControl Module Dimensions - 25.7 x 33.1 x 7.0 cmIncludes - 3 meter connection cable for Automated Thermal Cycler (ATC)
Quantity 1 cable
Volume (Metric) 10-100 μL
Weight 9.4 kg
Block Configurations 17.9 x 31.7 x 13.3 cm
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.