Learn More
United Scientific™ Conical Measuring Cylinders, Polypropylene
Unique design provides users with a different option than standard cylinders.
Supplier: United Scientific™ 81153
Description
- Our polypropylene conical measuring cylinders are ideal for measuring small quantities of reagents.
- They are autoclavable and have excellent contact clarity.
- The hexagonal base provides excellent stability to the cylinder, and the tapered spout enables easy pouring of reagents.
- Features easy-to-read raised graduations.
Specifications
| 125 mL | |
| 5 mL | |
| 4.76 in. | |
| Graduated Measuring Cylinder |
| Polypropylene | |
| Hexagonal | |
| 121 mm |