missing translation for 'onlineSavingsMsg'
Learn More
Learn More
United Scientific™ Conical Measuring Cylinders, Polypropylene
Unique design provides users with a different option than standard cylinders.
$31.79 - $86.82
Specifications
| Material | Polypropylene |
|---|---|
| Base Type | Hexagonal |
| Type | Graduated Measuring Cylinder |
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
|
S16003
|
United Scientific™
81154 |
200 mL |
Pack of 12 Each for $86.82
|
|
|||||
|
S16004
|
United Scientific™
81151* |
25 mL |
Pack of 12 Each for $35.68
|
|
|||||
|
S16055
|
United Scientific™
81150 |
12 mL |
Pack of 12 Each for $31.79
|
|
|||||
|
S16075
|
United Scientific™
81152 |
50 mL |
Pack of 12 Each for $46.93
|
|
|||||
|
S15792
|
United Scientific™
81153 |
125 mL |
Pack of 12 Each for $58.36
|
|
|||||
Description
- Our polypropylene conical measuring cylinders are ideal for measuring small quantities of reagents.
- They are autoclavable and have excellent contact clarity.
- The hexagonal base provides excellent stability to the cylinder, and the tapered spout enables easy pouring of reagents.
- Features easy-to-read raised graduations.
Specifications
| Polypropylene | |
| Hexagonal | |
| Graduated Measuring Cylinder |