missing translation for 'onlineSavingsMsg'
Learn More

Conductivity/TDS Standard, 84 μS/cm, Reagents

Catalog No. 01335516
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335516 1 L
01335515 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01335516 Supplier Labchem Inc Supplier No. CC9008401A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Conductivity/TDS Standard, 84 μS/cm, NIST Traceable, use for Conductivity meter calibration. Manufactured in ISO 9001 facility. Accredited to ISO 17025, NIST Traceable.

Specifications

Chemical Name or Material Conductivity/TDS Standard, 84 μS/cm
Packaging Poly Bottle
CAS 7732-18-5, 7447-40-7
Specific Gravity 1.0007
Physical Form Liquid
Molecular Formula KCl/H2O
Technique Calibration of Conductivity Instrument
Color Colorless
pH 5.4 to 8.6
Boiling Point 100°C
For Use With (Equipment) Conductivity Meter
Solubility Completely miscible
Quantity 1 L
Show More Show Less