missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Standard, 500 μS/cm at 25°C (240 ppm TDS as NaCl), Ricca Chemical
Supplier: Ricca Chemical Company 224132
Specifications
| Conductivity/TDS Standard | |
| Natural Poly Bottle | |
| Odorless | |
| Colorless | |
| 0°C | |
| Completely miscible | |
| Conductivity Standard | |
| 240ppm as NaCl |
| 500μS/cm (500μmho/cm) at 25°C | |
| 7647-14-5,7732-18-5 | |
| Liquid | |
| 6.7 to 7.3 | |
| 100°C | |
| Conductivity Standard | |
| 1 L |