missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Standard, 500 μS/cm at 25°C (240 ppm TDS as NaCl), Ricca Chemical

Catalog No. 224116
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
224116 500 mL
224132 1 L
22411 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 224116 Supplier Ricca Chemical Company Supplier No. 224116

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Conductivity/TDS Standard
Packaging Natural Poly Bottle
Name Note 500μS/cm (500μmho/cm) at 25°C
CAS 7647-14-5, 7732-18-5
Odor Odorless
Physical Form Liquid
Color Colorless
pH 6.7 to 7.3
Melting Point 0°C
Boiling Point 100°C
Solubility Completely miscible
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 500 mL
Concentration or Composition (by Analyte or Components) 240ppm as NaCl
Show More Show Less