missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Standard, 46.7 μS/cm at 25°C (22.0 ppm TDS as NaCl), Ricca Chemical

Supplier:  Ricca Chemical Company 22360732

 View more versions of this product

Catalog No. 223607-32

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Conductivity/TDS Standard
7647-14-5,7732-18-5
22.0ppm as NaCl
Liquid
100°C
Conductivity Standard
Natural Poly Bottle
98.0,0.0022
100.0,0.0022
Colorless
Conductivity Standard
1 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions