missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Standard, 46.7 μS/cm at 25°C (22.0 ppm TDS as NaCl), Ricca Chemical

Catalog No. 22360716
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22360716 500 mL
22360732 1 L
2236071 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22360716 Supplier Ricca Chemical Company Supplier No. 2236.0716
Only null left

Specifications

Chemical Name or Material Conductivity/TDS Standard
Packaging Natural Poly Bottle
CAS 7647-14-5, 7732-18-5
Concentration or Composition Notes 22.0ppm as NaCl
CAS Min % 98.0, 0.0022
CAS Max % 100.0, 0.0022
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 500 mL
Show More Show Less