missing translation for 'onlineSavingsMsg'
Learn More

Conductivity/TDS Standard, 1409 μS/cm, Reagents

Catalog No. 01335487
Change view
Click to view available options
Quantity:
1 L
4 L
10 L
20 L
200 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335487 1 L
01335488 10 L
01335486 200 L
01335485 20 L
01335484 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01335487 Supplier Labchem Inc Supplier No. CC9139001A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Conductivity/TDS Standard, 1409 μS/cm, NIST Traceable, use for Conductivity meter calibration. Manufactured in ISO 9001 facility. Accredited to ISO 17025, NIST Traceable.

Specifications

Chemical Name or Material Conductivity/TDS Standard, 1409 μS/cm
Packaging Poly Bottle
CAS 7732-18-5, 7447-40-7
Specific Gravity 1.0004
Physical Form Liquid
Molecular Formula KCl/H2O
Technique Calibration of Conductivity Instrument
Color Colorless
pH 5.4 to 8.6
Boiling Point 100°C
For Use With (Equipment) Conductivity Meter
Solubility Completely miscible
Quantity 1 L
Show More Show Less