missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Standard, 100,000 μS/cm (100 mS/cm) at 25°C, Ricca Chemical

Catalog No. 224932
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
224932 1 L
224916 500 mL
22491 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 224932 Supplier Ricca Chemical Company Supplier No. 224932
Only null left

65,750 ppm TDS as NaCl

Specifications

Chemical Name or Material Conductivity/TDS Standard
Packaging Natural Poly Bottle
Name Note 100mS/cm (100, 000μmho/cm) at 25°C
CAS 7647-14-5, 7732-18-5
CAS Min % 93.0, 4.85
CAS Max % 97.0, 5.05
Physical Form Liquid
Color Colorless
Melting Point Approximately 0°C
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 1 L
Concentration or Composition (by Analyte or Components) 65, 750ppm as NaCl
Show More Show Less