missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Standard, 100 μS/cm at 25°C (47.2 ppm TDS as NaCl), Ricca Chemical
Supplier: Ricca Chemical Company 223720
Specifications
| Conductivity/TDS Standard | |
| Pack | |
| 98.0,0.0046 | |
| Liquid | |
| 6.7 to 7.3 | |
| 100°C | |
| Conductivity Standard | |
| 20 x 20 mL |
| 100μS/cm (100μmho/cm) at 25°C | |
| 7647-14-5,7732-18-5 | |
| 100.0,0.0048 | |
| Colorless | |
| 0°C | |
| Completely miscible | |
| Conductivity Standard | |
| 47.2ppm as NaCl |