missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Standard, 100 μS/cm at 25°C (47.2 ppm TDS as NaCl), Ricca Chemical

Catalog No. 2237100P
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
20 x 20 mL
4 L
Packaging:
Natural Poly Bottle
Pack
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
2237100P 100 mL Pack
223720 20 x 20 mL Pack
223716 500 mL Natural Poly Bottle
223732 1 L Natural Poly Bottle
22371 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 2237100P Supplier Ricca Chemical Company Supplier No. 2237100P

Specifications

Chemical Name or Material Conductivity/TDS Standard
Packaging Pack
Name Note 100μS/cm (100μmho/cm) at 25°C
CAS 7647-14-5, 7732-18-5
CAS Min % 98.0, 0.0046
CAS Max % 100.0, 0.0048
Physical Form Liquid
Color Colorless
pH 6.7 to 7.3
Melting Point 0°C
Boiling Point 100°C
Solubility Completely miscible
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 100 mL
Concentration or Composition (by Analyte or Components) 47.2ppm as NaCl
Show More Show Less