missing translation for 'onlineSavingsMsg'
Learn More

Conductivity/TDS Standard, 10,000 μmho/cm (10 mS/cm), Reagents

Catalog No. 01335454
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
20 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335454 100 mL
01335453 1 L
01335452 20 L
01335451 4 L
01335450 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01335454 Supplier Labchem Inc Supplier No. CC970000100S

missing translation for 'validMainframeMsg'

Conductivity/TDS Standard, 10,000 μmho/cm (10 mS/cm), NIST Traceable, use for Conductivity meter calibration. Manufactured in ISO 9001 facility. Accredited to ISO 17025, NIST Traceable.

Specifications

Chemical Name or Material Conductivity/TDS Standard, 10,000 μmho/cm (10 mS/cm)
Packaging Poly Bottle
CAS 7732-18-5, 7447-40-7
Specific Gravity 1.004
Physical Form Liquid
Molecular Formula KCl/H2O
Technique Calibration of Conductivity Instrument
Color Colorless
pH 5.4 to 8.6
Boiling Point 100°C
For Use With (Equipment) Conductivity Meter
Solubility Completely miscible
Quantity 100 mL
Show More Show Less