missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Standard, METTLER TOLEDO
Description
- It ensures accurate, reliable measurements essential for quality control, research, and environmental monitoring.
- The convenient size supports multiple calibrations and easy handling, helping maintain device accuracy through stable calibration.
Specifications
Specifications
| Chemical Name or Material | Conductivity Standard 500,000 μS/cm at 25°C |
| Packaging | Bottle |
| CAS | 7647-01-0, 7732-18-5 |
| Conductivity | 500,000 μS/cm |
| Type | Conductivity Standard |
| Molecular Weight (g/mol) | Unknown |
| Quantity | 250 mL |
| Percent Purity | Unknown |
| Grade | Laboratory |