missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Standard, 84μmho/cm, LabChem™

Catalog No. LC187501
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC187501 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC187501 Supplier LabChem Supplier No. LC187501
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Conductivity Standard, 84μmho/cm
Packaging Poly Bottle
CAS 7447-40-7, 7732-18-5
CAS Max % 99.996, 0.004
Identification Passes Test
Density 1g/mL
Physical Form Liquid
Linear Formula KCl
Color Colorless
Solubility Soluble in water
Quantity 500 mL
Formula Weight 74.55
Grade Certified
Show More Show Less