missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Standard, 700 μS/cm at 25°C (342 ppm TDS as NaCl), Ricca Chemical
Conductivity/TDS Standard, 700 μS/cm (700 μmho/cm) at 25°C, 342 ppm as NaCl
$113.40 - $176.44
Specifications
| Chemical Name or Material | Conductivity/TDS Standard |
|---|---|
| Packaging | Natural Poly Bottle |
| CAS | 7647-14-5,7732-18-5 |
| CAS Min % | 98.0,0.03 |
| Concentration or Composition Notes | 342ppm as NaCl |
Specifications
| Conductivity/TDS Standard | |
| 7647-14-5,7732-18-5 | |
| 342ppm as NaCl | |
| Liquid | |
| 100°C | |
| Conductivity Standard |
| Natural Poly Bottle | |
| 98.0,0.03 | |
| 100.0,0.03 | |
| Colorless | |
| Conductivity Standard |