missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Standard, 70 μS/cm at 25°C (33.0 ppm TDS as NaCl), Ricca Chemical

Catalog No. p-3251565
Change view
Click to view available options
Quantity:
500 mL
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22361416 500 mL
2236141 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22361416 Supplier Ricca Chemical Company Supplier No. 2236.1416
Only null left

Specifications

Chemical Name or Material Conductivity/TDS Standard
Packaging Natural Poly Bottle
CAS 7647-14-5, 7732-18-5
Concentration or Composition Notes 33.0ppm as NaCl
CAS Min % 98.0, 0.0032
CAS Max % 100.0, 0.0034
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 500 mL
Show More Show Less