missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Standard, 70,000 μS/cm (70 mS/cm) at 25°C (44,000 ppm TDS as NaCl), Ricca Chemical
$107.44 - $165.66
Specifications
| Chemical Name or Material | Conductivity/TDS Standard |
|---|---|
| Packaging | Natural Poly Bottle |
| CAS | 7647-14-5,7732-18-5 |
| CAS Min % | 94.0,4.12 |
| Concentration or Composition Notes | 44,000ppm as NaCl |
Specifications
| Conductivity/TDS Standard | |
| 7647-14-5,7732-18-5 | |
| 44,000ppm as NaCl | |
| Liquid | |
| Approximately 0°C | |
| Conductivity Standard |
| Natural Poly Bottle | |
| 94.0,4.12 | |
| 98.0,4.29 | |
| Colorless | |
| Conductivity Standard |