missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Standard (500 μS/cm at 25°C, 2.5 KCl (T)), MilliporeSigma™ Supelco™

Catalog No. 111002566
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002566 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002566 Supplier Merck Emd Millipore Supplier No. 06999500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Conductivity standard can generally be used for the calibration of conductivity measurements. The conductivity standard solution used for conductivity measurements can be sodium chloride or potassium chloride solution of a definite concentration.

Featured Industries: Cleaning Products, Cosmetics, Flavors and Fragrances, Food and Beverages and Personal Care

Specifications

CAS 67-66-3
Density 0.995-1.014 g/cm3 (at 20°C)
Concentration ≤2.5% KCl (T)
Type Analytical Standard
Quantity 500 mL
Grade Analytical Standard