missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Standard, 5 μS/cm at 25°C, Ricca Chemical

Catalog No. 2236024
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2236024 120 mL
22360216 500 mL
22360232 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 2236024 Supplier Ricca Chemical Company Supplier No. 2236.024

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Conductivity Standard
Packaging Natural Poly Bottle
Name Note 5μS/cm (5μmho/cm) at 25°C
CAS 7647-14-5, 7732-18-5
CAS Min % 98.0, 2.0E-4
CAS Max % 100.0, 2.0E-4
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 120 mL
Show More Show Less