missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Standard, 5 μS/cm at 25°C, Ricca Chemical
Supplier: Ricca Chemical Company 22360232
Specifications
| Conductivity/TDS Standard | |
| Natural Poly Bottle | |
| 98.0,2.0E-4 | |
| 100°C | |
| Conductivity Standard |
| 5-17, 000 Micromhos/cm at 25° C/4.7 ppm TDS as NaCl | |
| 7647-14-5,7732-18-5 | |
| 100.0,2.0E-4 | |
| Conductivity Standard | |
| 1 L |