missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Standard (30000 μS/cm at 25°C), MilliporeSigma™ Supelco™

Catalog No. 111002564
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002564 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002564 Supplier Merck Emd Millipore Supplier No. 91916500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Conductivity standard may be used for the calibration of conductivity sensors for conductivity measurements. The conductivity standard solution used is sodium chloride or potassium chloride solution of a definite concentration.

Featured Industries: Cleaning Products, Cosmetics, Flavors and Fragrances, Food and Beverages and Personal Care

Specifications

CAS 67-66-3
Density 0.995-1.014 g/cm3 (at 20°C)
Concentration ≤2.5% KCl (T)
Type Analytical Standard
Quantity 500 mL
Grade Analytical Standard