missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Standard (3000 S/cm at 25°C), ≤2.5% KCl (T), MilliporeSigma™ Supelco™

Catalog No. 111002568
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002568 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002568 Supplier Merck Emd Millipore Supplier No. 44348500ML
Only null left

Conductivity standard can generally be used for the calibration of conductivity measurements. The conductivity standard solution, which can be used for conductivity measurements can be sodium chloride or potassium chloride solution of a definite concentration.

Cleaning Products, Cosmetics, Flavors and Fragrances, Food and Beverages and Personal Care

Specifications

CAS 67-66-3
Density 0.995 to 1.014 g/cm3 (at 20°C)
Type Analytical Standard
Quantity 500 mL
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) ≤2.5% KCl (T)