missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Standard, 3000μmho/cm, Certified, LabChem™
Specifications
Specifications
| Chemical Name or Material | Conductivity Standard |
| Packaging | Poly Bottle |
| Name Note | 3000μmho/cm |
| CAS | 7447-40-7, 7732-18-5 |
| CAS Max % | 99.84, 0.16 |
| Identification | Passes Test |
| Density | 1g/mL |
| Physical Form | Liquid |
| Linear Formula | KCl |
| Color | Colorless |
| Show More |