missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Standard (300 S/cm at 25°C), ≤2.5% KCl (T), MilliporeSigma™ Supelco™
Description
Cleaning Products, Cosmetics, Flavors and Fragrances, Food and Beverages and Personal Care
Specifications
Specifications
| CAS | 67-66-3 |
| Density | 0.995 to 1.014 g/cm3 (at 20°C) |
| Type | Analytical Standard |
| Quantity | 500 mL |
| Grade | Analytical Standard |
| Concentration or Composition (by Analyte or Components) | ≤2.5% KCl (T) |