missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride Conductivity Standard, 2767 μS/cm at 25°C, 0.02 M, Ricca Chemical

$169.70 - $228.50

Specifications

Chemical Name or Material Conductivity Standard
Packaging Natural Poly Bottle
pH Approximately 7
Boiling Point 100°C
Solution Type Conductivity Standard
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
588803-16
View Documents
Ricca Chemical Company
58880316
500 mL
Each for $169.70
Only null left
 
588803-32
View Documents
Ricca Chemical Company
58880332
1 L
Each for $228.50
Only null left
 
Description

Description

Specifications

Specifications

Conductivity Standard
Approximately 7
Conductivity Standard
Potassium Chloride in Water
Natural Poly Bottle
100°C
Conductivity Standard
Videos
SDS
Documents

Documents