missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Standard, 2765μmho/cm, Certified, LabChem™

Catalog No. LC187722
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC187722 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC187722 Supplier LabChem Supplier No. LC187722

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Conductivity Standard
Packaging Poly Bottle
Name Note 2765μmho/cm
CAS 7447-40-7, 7732-18-5
CAS Max % 99.85, 0.15
Identification Passes Test
Density 1g/mL
Physical Form Liquid
Linear Formula KCl
Color Colorless
Solubility Soluble in water
Quantity 1 L
Formula Weight 74.55
Grade Certified
Show More Show Less