missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Standard, 23.8 μS/cm at 25°C (11.2 ppm TDS as NaCl), Ricca Chemical
$109.09 - $203.03
Specifications
| Chemical Name or Material | Conductivity/TDS Standard |
|---|---|
| Packaging | Natural Poly Bottle |
| CAS | 7647-14-5,7732-18-5 |
| CAS Min % | 98.0,0.0011 |
| Concentration or Composition Notes | 11.2ppm as NaCl |
Specifications
| Conductivity/TDS Standard | |
| 7647-14-5,7732-18-5 | |
| 11.2ppm as NaCl | |
| Liquid | |
| 100°C | |
| Conductivity Standard |
| Natural Poly Bottle | |
| 98.0,0.0011 | |
| 100.0,0.0011 | |
| Colorless | |
| Conductivity Standard |