missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Standard (200 μS/cm at 25°C), MilliporeSigma™ Supelco™

Catalog No. 111002565
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002565 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002565 Supplier Merck Emd Millipore Supplier No. 06811500ML
Featured Industries: Cleaning Products, Cosmetics, Flavors and Fragrances, Food and Beverages and Personal Care

Specifications

CAS 67-66-3
Density 0.995-1.014 g/cm3 (at 20°C)
Concentration ≤2.5% KCl (T)
Type Analytical Standard
Quantity 500 mL
Grade Analytical Standard