missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Standard, 200,000 μS/cm (200 mS/cm) at 25°C, Ricca Chemical

Catalog No. 22492032
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22492032 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22492032 Supplier Ricca Chemical Company Supplier No. 2249.2032
Only null left

Specifications

Chemical Name or Material Conductivity Standard
Packaging Natural Poly Bottle
Name Note 200 mS/cm (200, 000 μmho/cm) at 25°C
CAS 7647-14-5, 7732-18-5
CAS Min % 83.0, 14.82
CAS Max % 87.0, 15.42
Physical Form Liquid
Color Colorless
pH 6.7 to 7.3
Boiling Point Approximately 100.1°C
For Use With (Equipment) Conductivity Meters and Probes
Solubility Miscible with water
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 1 L
Grade Laboratory
Show More Show Less