missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Standard, 20,000 μS/cm (20 mS/cm) at 25°C (11,000 ppm TDS as NaCl), Ricca Chemical

$120.80 - $190.63

Specifications

Chemical Name or Material Conductivity/TDS Standard
Packaging Natural Poly Bottle
CAS 7647-14-5,7732-18-5
Concentration or Composition Notes 11,000ppm as NaCl
Physical Form Liquid
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
2247-16
View Documents
Ricca Chemical Company
224716
500 mL
Each for $120.80
Only null left
 
2247-32
View Documents
Ricca Chemical Company
RI224732
1 L
Each for $190.63
Only null left
 
Description

Description

Specifications

Specifications

Conductivity/TDS Standard
7647-14-5,7732-18-5
Liquid
MFCD00003477
100°C
Conductivity Standard
sodium chloride
5234
Natural Poly Bottle
11,000ppm as NaCl
ClNa
Colorless
FAPWRFPIFSIZLT-UHFFFAOYSA-M
[Na+].[Cl-]
58.44
CHEBI:26710
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'