missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Standard, 2500 μS/cm at 25°C, Ricca Chemical

Catalog No. 22442532
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22442532 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22442532 Supplier Ricca Chemical Company Supplier No. 2244.2532
Only null left

Specifications

Chemical Name or Material Conductivity Standard
Packaging Natural Poly Bottle
CAS 7647-14-5, 7732-18-5
CAS Min % 98.0, 0.12
CAS Max % 100.0, 0.13
Physical Form Liquid
Color Colorless
pH 6.7 to 7.3
Melting Point 0.0°C
Boiling Point 100°C
For Use With (Equipment) Conductivity Meters and Probes
Solubility Miscible with water
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 1 L
Grade Laboratory
Show More Show Less