missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Standard, 16,630 μS/cm (16.63 mS/cm) at 25°C (9100 ppm TDS as NaCl), Ricca Chemical
$106.19 - $399.03
Specifications
| Chemical Name or Material | Conductivity/TDS Standard |
|---|---|
| Packaging | Natural Poly Bottle |
| CAS | 7647-14-5,7732-18-5 |
| CAS Min % | 97.0,0.92 |
| Concentration or Composition Notes | 9100ppm as NaCl |
Specifications
| Conductivity/TDS Standard | |
| 7647-14-5,7732-18-5 | |
| 9100ppm as NaCl | |
| Liquid | |
| 100°C | |
| Conductivity Standard |
| Natural Poly Bottle | |
| 97.0,0.92 | |
| 100.0,0.96 | |
| Colorless | |
| Conductivity Standard |